missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PIBF1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5559
Additional Details : Weight : 0.00970kg
Description
PIBF1 Polyclonal specifically detects PIBF1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PIBF1 | |
Polyclonal | |
Unconjugated | |
Q8WXW3 | |
PIBF1 | |
E. coli-derived recombinant human PIBF1 Glu19-Ala419 Accession # Q8WXW3 | |
100 μg | |
Primary | |
Detects human PIBF1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
C13orf24, chromosome 13 open reading frame 24, KIAA1008, PIBF, progesterone immunomodulatory binding factor 1, progesterone-induced blocking factor 1, progesterone-induced-blocking factor 1, RP11-505F3.1 | |
Sheep | |
Affinity Purified | |
RUO | |
10464 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |