missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PLA2G2A Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5374
This item is not returnable.
View return policy
Description
PLA2G2A Polyclonal specifically detects PLA2G2A in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PLA2G2A | |
Polyclonal | |
Unconjugated | |
P14555 | |
PLA2G2A | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PLA2G2A Asn21-Cys144 Accession # P14555 | |
100 μg | |
Primary | |
Detects human PLA2G2A in direct ELISAs and Western blots. In Western blots, approximately 60% cross-reactivity with recombinant mouse PLA2G2A is observed and less than 5% cross-reactivity with recombinant human (rh) PLA2G1B and rhPLA2G7 is observed. | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
EC 3.1.1.4, GIIC sPLA2, Group IIA phospholipase A2, MOM1, Non-pancreatic secretory phospholipase A2, NPS-PLA2, Phosphatidylcholine 2-acylhydrolase 2A, phospholipase A2, group IIA (platelets, synovial fluid), phospholipase A2, membrane associated, PLA2B, PLA2BPLA2, PLA2L, PLA2LPLA2S, PLAS1, RASF-A, sPLA2 | |
Sheep | |
Affinity Purified | |
RUO | |
5320 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only