Learn More
Gibco™ Human Plasma-Like Medium (HPLM)
Gibco Human Plasma-like Medium (HPLM) is designed to resemble the natural cellular environment found in the body, mimicking the metabolic profile of human plasma.
Brand:  Gibco™ A4899101
57.48 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 17100059
  • £60.50 / 500mL
In Stock
to see stock.
Description
Description
  • Physiologically relevant—designed to resemble the natural cellular environment found in the body; formulated with over than 60 polar metabolites and plasma-like salt concentrations
  • Peer reviewed—extensive publication of research that used HPLM
  • Easy to use—direct replacement for conventional media when supplemented with FBS
The widely used classic synthetic cell culture media, including MEM, DMEM, RPMI 1640, and DMEM/F-12 contain glucose, amino acids, vitamins, and salts at concentrations that often do not reflect those found in human plasma. These media also lack additional plasma components needed to mimic the metabolic profile of human plasma. When studying cancer and other diseases, results with more physiological relevance will enable researchers to improve their understanding of human function and illness.
The HPLM solution
HPLM contains more than 60 polar metabolites such as amino acids, nucleic acids, sugars, and small organic acids at concentrations found in human plasma. The salt concentrations in HPLM mimic the salt concentrations found in human plasma. In resembling the natural cellular environment found in the body, HPLM provides researchers with the ability to study the impact of physiologically relevant cell media on their specific applications.
When supplemented with fetal bovine serum (FBS), HPLM is capable of supporting cell growth and viability comparable to those of conventional basal media supplemented with FBS. For most cell lines, adaptation is not required to transition from conventional medium to HPLM.
The complete HPLM formulation is available (hyperlink to technical-resource page with complete HPLM formulation).
cGMP manufacturing and quality system
HPLM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
Mammalian Cells
Liquid
Standard Serum Supplementation
Glutamine, Phenol Red
Mammalian Cell Culture
Mammalian Cell Culture
Human Plasma-Like Medium
Sterile
No HEPES
500 mL