missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLUNC ELISA Kit
Human PLUNC quantitates human PLUNC in serum, plasma, supernatant.
Brand: Invitrogen™ EH372RB
This item is not returnable.
View return policy
Description
Human PLUNC quantitates human PLUNC in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PLUNC.
PLUNC is the human homolog of murine plunc and is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this protein is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. This gene is the human homolog of murine plunc, and like the mouse gene, is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this gene is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. Multiple transcript variants resulting from alternative splicing in the 3' UTR have been detected, but the full-length nature of only two is known.Specifications
bA49G10.5; BPI fold containing family A member 1; BPI fold containing family A, member 1; BPI fold-containing family A member 1; Bpifa1; ES cell-related protein; ligand-binding protein RYA3; LPLUNC3; lung-specific protein X; LUNX; LUNXNASG; NASG; Nasopharyngeal carcinoma-related protein; palate lung and nasal epithelium clone protein; palate, lung and nasal epithelium associated; palate, lung, and nasal epithelium associated; palate, lung, and nasal epithelium carcinoma associated protein; palate, lung, and nasal epithelium clone; palate, lung, and nasal epithelium expressed transcript; Plunc; protein Plunc; secretory protein in upper respiratory tracts; short palate; Short PLUNC1; SPLUNC1; SPLUNC1SPURT; SPURT; tracheal epithelium enriched protein; tracheal epithelium-enriched protein; UNQ787/PRO1606; Von Ebner protein Hl | |
40 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Bpifa1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
40 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q9NP55 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
PLUNC | |
4 hr 45 min |