Learn More
STEMCELL Technologies Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
For characterization of hPSC-derived progenitor cells of definitive endoderm, hepatic, intestinal, or pancreatic lineages
Brand:  STEMCELL Technologies 07533
Encompass_Preferred
 View more versions of this product
Product Code. 17159291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • The Human Pluripotent Stem Cell (hPSC)-Derived Endoderm Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of hPSC-derived definitive endodermal progenitor cells and their differentiated progeny including pancreatic, hepatic, and intestinal cells
  • This qPCR array provides the gene expression profile of hPSC-derived endodermal cells following in vitro differentiation, and can aid in development of protocols for directed differentiation towards endodermal lineages
  • Genes were selected based on their demonstrated expression in hPSCs, or in hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (D'Amour et al, including pancreatic (Kroon et al, hepatic (Hay et al, and intestinal progenitor cells (Grapin-Botton & Melton)
Specifications
Specifications
Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Pancreatic Cells; Pluripotent Stem Cells
96-well Plate Type 3
Real Time PCR (qPCR)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Pluripotent Stem Cell-Derived Endoderm qPCR Array