missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pluripotent Stem Cell Functional Identification Kit

Verify pluripotency in human stem cells by in vitro functional differentiation

Brand:  R&D Systems™ SC027B

511.43 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 30001925

  • £650.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Verifies the pluripotency of your starting population
  • Only takes 5 days
  • Reduces experimental variation
Specifications

Specifications

  • Differentiation Base Media Supplement (50X)
  • Ectoderm, Mesoderm, and Endoderm Differentiation Supplements
  • Ectoderm Marker: Goat Anti-Human Otx2 Antigen Affinity-purified Polyclonal Antibody
  • Mesoderm Marker: Goat Anti-Human Brachyury Antigen Affinity-purified Polyclonal AntibodyV
  • Endoderm Marker: Goat Anti-Human SOX17 Antigen Affinity-purified Polyclonal Antibody
The quantity of each component in this kit is sufficient to make 200 mL of media for differentiation
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Human Pluripotent Stem Cell Functional Identification Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers