missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PP2A Catalytic Subunit Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB16531
This item is not returnable.
View return policy
Description
PP2A alpha Monoclonal specifically detects PP2A alpha in Human samples. It is validated for Immunohistochemistry.Specifications
PP2A alpha | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P67775 | |
PPP2CA | |
E. coli-derived recombinant human PP2A-alpha Catalytic Subunit Trp209-Leu309 Accession # P67775 | |
100 μg | |
Primary | |
Detects human PP2A Catalytic Subunit in direct ELISAs. Because PP2A-alpha and PP2A-beta Catalytic Subunits are identical in the region used for the antigen, it is expected that both isoforms will be detected by the antibody. | |
Human | |
Purified |
Immunohistochemistry | |
234532 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, PPP2CA, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5515 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only