missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro Relaxin-3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£146.86 - £333.68
Specifications
Antigen | Relaxin-3 |
---|---|
Clone | 332112 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732847
|
R&D Systems
MAB3107-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18181151
|
R&D Systems
MAB3107 |
100 μg |
£333.68
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot.Specifications
Relaxin-3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8WXF3 | |
117579 | |
E. coli-derived recombinant human Relaxin-3 Arg26-Cys142 Accession # Q8WXF3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Pro Relaxin-3 in direct ELISAs and Western blots. No cross-reactivity with mature human Relaxin-3, human Pro Relaxin-1 or human Pro Relaxin-2 is observed. |
332112 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
H3, INSL7, M3, relaxin 3, Relaxin3, RLN3, RXN3, ZINS4 | |
RLN3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only