Learn More
Novus Biologicals™ Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric)
Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: Novus Biologicals™ NBP2-76465
This item is not returnable.
View return policy
Description
The Human, Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) measures human Procollagen Type 1 N-Terminal Propeptide in serum, plasma and other biological fluids.Specifications
15.63 to 1000 pg/mL | |
ELISA Kit (Colorimetric) | |
The Human Procollagen Type 1 N-Terminal Propeptide ELISA Kit (Colorimetric) recognizes Human P1NP in samples. No significant cross-reactivity or interference between Human P1NP and analogues was observed. | |
Human | |
Cellular Markers, Extracellular Matrix, Metastasis | |
amino-terminal propeptide, PINP |
9.38 pg/mL | |
Biological Fluids, Plasma, Serum | |
1277 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |