Learn More
Abnova™ Human PRRX2 Partial ORF (NP_057391.1, 151 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£280.00 - £425.00
Specifications
Accession Number | NP_057391.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51450 |
Molecular Weight (g/mol) | 37.07kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085515
|
Abnova™
H00051450-Q02.25UG |
25 ug |
£425.00
25µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075515
|
Abnova™
H00051450-Q02.10UG |
10 ug |
£280.00
10µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins. Expression is localized to proliferating fetal fibroblasts and the developing dermal layer, with downregulated expression in adult skin. Increases in expression of this gene during fetal but not adult wound healing suggest a possible role in mechanisms that control mammalian dermal regeneration and prevent formation of scar response to wounding. The expression patterns provide evidence consistent with a role in fetal skin development and a possible role in cellular proliferation. [provided by RefSeq]
Sequence: WFQNRRAKFRRNERAMLASRSASLLKSYSQEAAIEQPVAPRPTALSPDYLSWTASSPYSTVPPYSPGSSGPATPGVNMANSIASLRLKAKEFSLHHSQVPTVNSpecifications
NP_057391.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC19843/PMX2/PRX2 | |
PRRX2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51450 | |
PRRX2 (Human) Recombinant Protein (Q02) | |
WFQNRRAKFRRNERAMLASRSASLLKSYSQEAAIEQPVAPRPTALSPDYLSWTASSPYSTVPPYSPGSSGPATPGVNMANSIASLRLKAKEFSLHHSQVPTVN | |
RUO | |
PRRX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |