missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant IL-15, ACF

Product Code. 17189871
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
This item is not returnable. View return policy

Product Code. 17189871

Brand: STEMCELL Technologies 78218.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 15, animal component-free

  • Interleukin 15 (IL-15) is a four-alpha helix bundle cytokine with many similar properties to IL-2
  • The IL-15 receptor is a heterotrimeric receptor composed of IL-15Ra (the high-affinity receptor for IL-15), as well as IL-2/15Rb (CD122) and common gamma chain (CD132)
  • IL-15 binds to IL-15Rα receptor and can then be presented in trans to IL-2/15Rb and common gamma chain on other cells
  • Trans-presentation is thought to be the major mechanism by which IL-15-mediated responses occur in mice, although may not be necessary in humans (Castillo et al.) The cytoplasmic domains of IL-2/15Rb and common gamma chain mediate signaling to activate JAK/STAT and PI3K pathways
  • IL-15 supports the survival and proliferation of naïve CD4+ and CD8+ T cells, and promotes homeostasis of memory T cells
  • IL-15 also promotes the survival and differentiation of NK cells and regulates their cytolytic activity (Ma et al.) This product is animal component-free

Specifications

Name IL-T, Interleukin-15
Quantity 100 μg
Cell Type Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.