missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-2, ACF
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor
- It binds to heterotrimeric receptors consisting of CD25, CD122, and CD132
- Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival (Ma et al.) The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts
- IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ T regulatory cells and affects development of follicular helper T cells
- IL-2 also controls inflammation by inhibiting Th17 differentiation
- This product is animal component-free
Specifications
Specifications
| Name | Aldesleukin, Interleukin-2, T cell growth factor, TCGF |
| Quantity | 10 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+ |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction