missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RNF175 (aa 9-46) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP96683
This item is not returnable.
View return policy
Description
RNF175 gene ontology annotations related to this gene include endoplasmic reticulum unfolded protein response; ubiquitin-dependent ERAD pathway.Specifications
Q8N4F7 | |
Blocking Assay, Control | |
285533 | |
100 μL | |
RUO | |
RNF175 | |
Human | |
KAAPVLEAPPQQEQLSHTKLSAEDTWNLQQERMYKMHR | |
Liquid |
≥5.0 mg/mL | |
1M urea, PBS with no preservative; pH 7.4 | |
RNF175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
RING finger protein 175; RNF175 | |
Unconjugated | |
Recombinant | |
E. coli |