Learn More
Invitrogen™ APO-1/FAS (Soluble) Human ELISA Kit
For the quantitative detection of human sAPO-1/Fas.
Brand: Invitrogen™ BMS245TEN
2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes:
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sAPO-1/Fas
- Biotin-Conjugate anti-human sAPO-1/Fas monoclonal antibody
- Streptavidin-HRP
- Human sAPO-1/Fas Standard lyophilized, 2ng/mL upon reconstitution
- Sample Diluent
- Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
- Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
- Substrate Solution (tetramethyl-benzidine)
- Stop Solution (1M Phosphoric acid)
- Adhesive Films
Description
The Human Soluble APO-1/Fas ELISA quantitates Hu sAPO-1/Fas in human serum, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu sAPO-1/Fas. Principle of the method The Human APO-1/Fas solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
FAS (Fas, CD95, APO-1) is a 46 kDa transmembrane glycoprotein that functions as a cell death receptor of the TNFR (tumor necrosis factor receptor) superfamily. FAS also exists in a soluble form that weight approximately 26kDa. Stimulation of FAS results in aggregation of its intracellular FAS-associated death domains (FADD), formation of the death-inducing signaling complex (DISC) and activation of caspases. The auto-proteolytic processing of the caspases in the complex trigger a downstream cascade that leads to apoptosis. FAS has been shown to activate NF-kappaB, MAPK3/ERK1, MAPK8/JNK, and is involved in transducing the proliferating signals in normal diploid fibroblast and T cells. At least eight alternatively spliced transcript variants encoding seven distinct isoforms have been described. The isoforms lacking the transmembrane domain may negatively regulate the apoptosis mediated by the full-length isoform. The Fas/Fas ligand system has been shown to play a role in a number of human diseases such as AIDS, hepatitis and cancer. It is also assumed that induction of apoptosis through FAS ligand pathway is predominantly involved in anti-viral immune responses.Specifications
P25445 | |
13.2 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
ALPS1A,APO-1,APT1,CD951,FASTM,TNFRSF6,FAS | |
4.5% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
2 hr. 10 min. |
15.6-1,000 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
355 | |
3.1% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 10 μL; Supernatant, 10 μL | |
CD95, Fas receptor, APT 1, ALPS 1 A, TNFRSF6 | |
1 hr. 20 min. |
For Research Use Only