missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD44var (v5) (Soluble) Human ELISA Kit

For the quantitative detection of human sCD44var (v5).

Brand:  Invitrogen™ BMS220

 View more versions of this product

Product Code. 15541717

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes:
  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var (v5)
  • HRP-Conjugate anti-human sCD44var (v5) monoclonal antibody
  • Human sCD44var (v5) Standard lyophilized, 20ng/mL upon reconstitution
  • Sample Diluent
  • Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
  • Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Blue-Dye
  • Green-Dye
  • Adhesive Films
This item is not returnable. View return policy

Description

Description

The Human CD44var (v5) ELISA quantitates Hu CD44var (v5) in human serum, plasma, urine, amniotic fluid or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CD44var (v5). Principle of the method The Human CD44var (v5) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration. It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). This protein participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full length nature of some of these variants has not been determined. Alternative splicing is the basis for the structural and functional diversity of this protein, and may be related to tumor metastasis. [provided by RefSeq, Jul 2008]
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P16070
0.14 ng/mL
ELISA Kit
Human
Colorimetric Microplate Reader
CDW44,CSPG8,ECMR-III,HCELL,HUTCH-I,IN,LHR,MC56,MDU2,MDU3,MIC4,Pgp1
3.6%
HRP
RUO
2°C to 8°C
Human
3 hr. 10 min.
0.31-10.0 ng/mL
HRP
Amniotic Fluid, Plasma, Serum, Supernatant, Urine
ELISA
960
5.8%
Pre-coated 96 well plates, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Amniotic Fluid, 20 μL; Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL; Urine,20 μL
VFF-8 Phagocytic glycoprotein I, Hermes antigen
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only