missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Human Serum, Pooled
Used as a media component to neutralize cell free virus.
Brand: MP Biomedicals™ 092930154
This item is not returnable.
View return policy
Description
Human pooled serum also used as a reagent in liposome binding assay and in preparation of sperm. This serum is obtained from pooled, normal clotted human blood.

Specifications
Golden Brown | |
Human Serum, Pooled | |
500 mL | |
Sterile | |
7 to 8 |
Store at -20°C | |
Liquid | |
5.5 to 7 g/dL | |
0.1 μm filtration |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Human Serum, Pooled > 500mL
Spot an opportunity for improvement?Share a Content Correction