missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SMO Partial ORF (NP_005622.1, 461 a.a. - 519 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006608-Q03.25ug
Detalles adicionales : Weight : 0.02000kg
Descripción
Sequence: GFVLITFSCHFYDFFNQAEWERSFRDYVLCQANVTIGLPTKQPIPDCEIKNRPSLLVEKEspecificaciones
NP_005622.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.12kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GFVLITFSCHFYDFFNQAEWERSFRDYVLCQANVTIGLPTKQPIPDCEIKNRPSLLVEK | |
RUO | |
SMO | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6608 | |
SMO (Human) Recombinant Protein (Q03) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Gx/SMOH | |
SMO | |
Recombinant | |
wheat germ expression system |