missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human Soluble Protein Master Buffer Kit
Along with Human soluble protein flex set system buffer kit employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
£515.00 - £2100.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Product Type | Cytometric Bead Array (CBA) Assay |
Validated Application | RUO |
Product Code | Brand | Includes | No. of Tests | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Tests | Price | Quantity & Availability | ||||
15839268
|
BD
558264 |
1 Bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 5mL capture bead diluent for Serum/Plasma, 1 bottle 130mL wash Buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial 1.0mL instrument setup bead F1, 1 vial 0.25mL instrument setup bead F9, 1 vial, 0.25mL PE instrument setup bead F1, 1 vial 0.5mL PE positive control detector | 100 |
£515.00
1 set |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15849268
|
BD
558265 |
1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 30mL capture bead diluent for Serum/Plasma, 1 bottle 650mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument | 500 |
£2100.00
1 set |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Buffers and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures or 500 samples and 10 instrument setup procedures
- Has been optimized for use with the CBA Human Soluble Protein Flex Sets
- Working assay range for most analytes in this system is 10 to 2,500pg/mL
Specifications
2°C to 8°C | |
Cytometric Bead Array (CBA) Assay | |
RUO |