missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SREC-I/SCARF1 ELISA Kit
Human SREC-I/SCARF1 quantitates human SREC-I/SCARF1 in serum, plasma, supernatant.
Brand: Invitrogen™ EH437RB
This item is not returnable.
View return policy
Description
Human SREC-I/SCARF1 quantitates human SREC-I/SCARF1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human SREC-I/SCARF1.
The protein encoded by this gene is a scavenger receptor that is expressed in endothelial cells and mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). Transcription factors ZNF444/EZF-2 and SP1 are found to be responsible for the regulation of this gene. Five alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
AA986099; acetyl LDL receptor; endothelial cells scavenger receptor; Kiaa0149; MGC47738; mKIAA0149; SCARF1; scavenger receptor class F member 1; scavenger receptor class F, member 1; scavenger receptor expressed by endothelial cells 1; Scavenger receptor expressed by endothelial cells 1 protein; Srec; SREC1; SREC-I | |
0.08 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Scarf1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.08 to 20 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q14162 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL | |
SCARF1 | |
4 hr 45 min |