Learn More
Invitrogen™ VAP-1 Human ELISA Kit
For the quantitative detection of human sVAP-1.
Brand: Invitrogen™ BMS259TEN
2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes:
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sVAP-1
- Biotin-Conjugate anti-human sVAP-1 monoclonal antibody
- Streptavidin-HRP
- Human sVAP-1 Standard lyophilized, 4ng/mL upon reconstitution
- Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
- Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
- Substrate Solution (tetramethyl-benzidine)
- Stop Solution (1M Phosphoric acid)
- Blue-Dye
- Green-Dye
- Red-Dye
- Adhesive Films
Description
The Human Vascular Adhesion Protein (Hu VAP-1) ELISA quantitates Hu VAP-1 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu VAP-1. Principle of the method The Human VAP-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.Specifications
Q16853 | |
19 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
HPAO,SSAO,VAP-1,VAP1 | |
3% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
31.3-2,000 pg/mL | |
Biotin | |
Amniotic Fluid, Plasma, Serum, Supernatant | |
ELISA | |
8639 | |
4.5% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers | |
10 x 96 Tests | |
Amniotic Fluid, 10 μL; Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL | |
Vascular adhesion protein 1, copper-containing 3 | |
1 hr. 20 min. |
For Research Use Only