missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TEP1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0474
This item is not returnable.
View return policy
Description
TP1 Synthetic Peptide, PEP-0474, from Invitrogen.
This peptide corresponds to 20 amino acids near the amino terminus of human TP1PEP-0474 can be used as a blocking peptide with polyclonal antibody PA5-20354.Specifications
7011 | |
purified | |
Human TEP1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q99973 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TEP1 | |
0.02% sodium azide | |
50 μg | |
PBS |