Learn More
Invitrogen™ Human TGF-beta 1 Recombinant Protein
Recombinant Human Transforming Growth Factor β 1 (TGFB1) is a bioactive protein intended for use in cell culture applications. TGFB1 is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types.
Brand: Invitrogen™ PHG9204
Description
Human TGF-beta 1 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
TGFB1 acts synergistically with TGF-α in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB1 activation and signaling may result in apoptosis.
- High purity—no interference from other proteins or contaminants
- High biological activity—more results with less protein
- Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High Purity Recombinant Bioactive Protein
Recombinant Human Transforming Growth Factor β 1 is one of more than 250 Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Transforming Growth Factor β 1 (TGFB1) specific activity is determined by measuring the TGFB1 dose-dependent inhibition of proliferation of mouse HT-2 cells stimulated with mouse IL-4.
Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
Specifications
Cell Culture | |
25 kDa | |
5μg | |
Human | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Oncology, Stem Cell Research, Cardiovascular Research, Differentiation, Bone Research | |
98 % |
7040 | |
Sequential Chromatography | |
TGF-β1 | |
Store in freezer (-5 to -30°C). | |
5 μg | |
Mammalian | |
Recombinant, Ligand | |
TGFs (Transforming Growth Factors) | |
Gibco™ |