missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TNF-alpha, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£79.00 - £1316.00
Specifications
Accession Number | P01375 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 7124 |
Molecular Weight (g/mol) | 17.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17853293
|
Gibco™
AF-300-01A-10UG |
10 μg |
£79.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17893793
|
Gibco™
AF-300-01A-100UG |
100 μg |
£270.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17843553
|
Gibco™
AF-300-01A-1MG |
1 mg |
£1316.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17833463
|
Gibco™
AF-300-01A-500UG |
500 μg |
£792.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.Specifications
P01375 | |
protein with no preservative | |
17.4 | |
E. coli | |
RUO | |
APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1 F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
7124 | |
Human TNF-alpha, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
Tnf | |
Unconjugated | |
VRSSSRTPSD KPVAHVVANP QAEGQLQWLN RRANALLANG VELRDNQLVV PSEGLYLIYS QVLFKGQGCP STHVLLTHTI SRIAVSYQTK VNLLSAIKSP CQRETPEGAE AKPWYEPIYL GGVFQLEKGD RLSAEINRPD YLDFAESGQV YFGIIAL | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |