missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TNF-alpha Recombinant Protein

Recombinant Human Tumor Necrosis Factor-α (TNF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC3013

 View more versions of this product

Product Code. 10542435

  • £1334.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Human TNF-alpha Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent cytotoxic effect on L929 cells in the presence of actinomycin D
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture
17.5 kDa
1 mg
Human
Carrier-Free
1 mg
E. coli
Recombinant, Ligand
TNF (Tumor Necrosis Factor)
Gibco™
7124
Sequential Chromatography
TNF-α
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Wet Ice
Cytokines & Receptors
Lyophilized
Immunology, Signal Transduction, Stem Cell Research, Inflammation, Oncology
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.