missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Torc2 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1590
Code : NEW
Additional Details : Weight : 0.01000kg
Description
CRTC2 Synthetic Peptide, PEP-1590, from Invitrogen.
This peptide corresponds to 16 amino acid peptide near the carboxy terminus of human CRTC2.PEP-1590 can be used as a blocking peptide with polyclonal antibody PA5-34547. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37ºC.Specifications
200186 | |
purified | |
Human Torc2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q53ET0 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
CRTC2 | |
0.02% sodium azide | |
50 μg | |
PBS |