missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total p27/Kip1 DuoSet IC ELISA
Human Total p27/Kip1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DYC2256-2
This item is not returnable.
View return policy
Description
The Human Total p27/Kip1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human p27/Kip1.Specifications
ELISA Development Kit for Intracellular Proteins | |
Human | |
62.50 to 4000 pg/mL | |
ELISA | |
2 Plates |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
1027 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |