missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human TSP-1 Platinum ELISA Kit
For the Detection of human Thrombospondin-1 (TSP-1)
Brand: Invitrogen™ BMS2100TEN
2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human TSP-1
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
0.044ng/mL | |
Human | |
10 x 96 Tests | |
TSP-1 |
Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
Thrombospondin 1, THBS1, THBS, THBS-1, TSP, TSP-1, TSP1 |