missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human WIF-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£146.86 - £442.75
Specifications
Antigen | WIF-1 |
---|---|
Clone | 133009 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729628
|
R&D Systems
MAB1342-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18140501
|
R&D Systems
MAB1342 |
500 μg |
£442.75
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WIF-1 Monoclonal specifically detects WIF-1 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
WIF-1 | |
LYOPH | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
AAD25402 | |
11197 | |
Mouse myeloma cell line NS0-derived recombinant human WIF-1 Gly29-Trp379 Accession # AAD25402 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human WIF-1 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse WIF-1 or recombinant human Wnt-7a is observed. |
133009 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
WIF1, WIF-1, WNT inhibitory factor 1 | |
WIF1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only