missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IGF-I protein

HumanKine™ recombinant protein

£42.00 - £133.00

Specifications

Accession Number P05019
Formulation 1 x PBS pH 7.4
Gene ID (Entrez) 3479
Molecular Weight (g/mol) 10 kDa
Source HEK293
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17600097
View Documents Promotion Details
Proteintech
HZ-1322-10UG
10 μg
£42.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17680087
View Documents Promotion Details
Proteintech
HZ-1322-1000UG
1000 μg
£133.00
1000µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17690087
View Documents Promotion Details
Proteintech
HZ-1322-100UG
100 μg
£45.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Insulin like growth factor-1, also known as Somatomedin-C is a growth factor which is structurally related to insulin and is an important regulator of growth and differentiation in various tissues and cell systems. Human IGF-1 is synthesized as two precursor isoforms with N- and alternate C-terminal propeptide. The two precursor isoforms are differentially expressed by various tissues. The proteolytic cleavage of the N- and C-terminal regions results in the mature IGF-1 protein which is identical between isoforms. IGF-1 binds to IGF-1 receptor and induces receptor autophosphorylation. This further phosphorylates Insulin receptor substrate -1 (IRS-1) and activates various downstream signaling pathways including the PI3-AKT, MAPK etc. (PMID: 17354613, 17113337, 29535161)
Specifications

Specifications

P05019
3479
HEK293
<1 EU/μg
Recombinant
2-14 ng/mL EC50
>95%
1 x PBS pH 7.4
10 kDa
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Human
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications