missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human IL-4 Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
£70.00 - £1782.00
Specifications
Gene ID (Entrez) | 3565 |
---|---|
Molecular Weight (g/mol) | 14 and 19 kDa, monomer, glycosylated |
Species | Human |
Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16841266
|
Proteintech™
HZ-1004-10UG |
10 μg |
£70.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16831266
|
Proteintech™
HZ-1004-100UG |
100 μg |
£358.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16821266
|
Proteintech™
HZ-1004-1000UG |
1000 μg |
£1782.00
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
3565 | |
Human | |
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 95% |
14 and 19 kDa, monomer, glycosylated | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Typically ≤ 0.6 ng/mL EC50 | |
Lyophilized |