Learn More
Proteintech™ HumanKine™ Recombinant Human LIF Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Brand:  Proteintech™ HZ-1292-10UG
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16891406
  • £65.00 / 10µg
Estimated Shipment: 15-08-2024
to see stock.
Description
Description
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Specifications
3976
10 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 1 ng/mL EC50
Lyophilized
35 to 53 kDa, glycosylated
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%