Learn More
Humic acid sodium salt, tech. 50-60% (as humic acid), Thermo Scientific Chemicals
Description
Humic acid sodium salt is a principal component of humic substances. It boosts plant growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >300°C |
Color | Black |
Merck Index | 14,4752 |
Solubility Information | Soluble in water (33.3 mg in 1 mL) |
Percent Purity | 50 to 60% |
Grade | Technical |
Quantity | 100 g |
Physical Form | Powder |
Assay | (as humic acid) |
Chemical Name or Material | Humic acid sodium salt |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M06L035 | Certificate of Analysis | 10/01/2025 | 041748.22,041748.36 |
X07K017 | Certificate of Analysis | 22/10/2024 | 041748.22,041748.36 |
W24K036 | Certificate of Analysis | 05/10/2024 | 041748.22,041748.36 |
U08K015 | Certificate of Analysis | 14/08/2024 | 041748.22,041748.36 |
N05K045 | Certificate of Analysis | 16/02/2024 | 041748.22,041748.36 |
Safety and Handling

- Humic acid, sodium salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Humic acids, sodiumsalts
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.