Learn More
Humic acid, Thermo Scientific Chemicals
Description
Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Solubility Information | Slightly soluble in water,with much swellingSoluble in organic solvents like acetone and benzenol. Slightly soluble in water. |
Quantity | 25 g |
Physical Form | Crystalline Powder |
Chemical Name or Material | Humic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.