missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testo™ Humidity Probe
Robust humidity probe measures up to 125°C, short-term up to 140°C
Brand: Testo™ 0636 2161
This item is not returnable.
View return policy
Description
Robust humidity probe for measuring exhaust ducts, and for measuring of material equilibrium moistureSpecifications
Probe diameter: 11.938 mm | |
±2% RH (+2 to +98% RH) | |
Humidity: 0 to 100%, Temperature: -20 to 125°C | |
299.974 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction