missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuR Monoclonal Antibody (3A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1167
Code : NEW
Additional Details : Weight : 0.00010kg
Description
HuR Monoclonal antibody specifically detects HuR in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot.Specifications
HuR | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.2-7.4 | |
B5DF91, P70372, Q15717 | |
ELAVL1 | |
Recombinant human HuR protein | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot, Western Blot | |
3A2 | |
Unconjugated | |
ELAVL1 | |
2410055N02Rik; ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; ELAV like RNA binding protein 1; ELAV like RNA binding protein 1a; ELAV1; Elavl1; elavl1 protein; elavl1a; elav-like generic protein; ELAV-like protein 1; ELAV-like protein 1; ELAV-like protein 1a; elav-related A; Elra; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-related A; etID309757.2; fa25f08; fb59a02; fc60g02; Hu antigen R; Hua; HU-antigen A; hu-antigen R; Human antigen R; HuR; melG; ribonucleoprotein; W91709; wu:fa25f08; wu:fb59a02; wu:fc60g02; zHuA | |
Mouse | |
Protein A | |
RUO | |
15568, 1994, 363854 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |