Learn More
Hamilton™ HVC Standard Ceramic Valves
Designed to provide superior performance with salt solutions.
Brand: Hamilton™ 8063-01
This item is not returnable.
View return policy
Description
- Rigid ceramic fluid path is resistant to scratching from salt crystals
- Provides superior life and performance
- Exceptionally easy to service and modify
- Maximizes flexibility and value
- Ideal for low pressure applications
Specifications
T 3-2 Model, 3 Ports, Aluminum Housing | |
Aluminum | |
Ceramic HV Valve |
PSD/4 and 8 Syringe Drive Modules | |
T 3-2 Model |