Learn More
Hamilton™ HVDP 2-5 Valve for Purge and Trap Syringes
For use with Hamilton Purge and Trap Syringes.
Brand: Hamilton™ 86789
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- With female luer
- 2-5 Model
- Aids in the analysis of drinking water samples according to EPA purge and trap concentration techniques
Specifications
Female Luer | |
Black |