missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HVEM/TNFRSF14 Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHTNFRSF14X10

Product Code. 17788033

  • £4370.00 / Pack of 10
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Q92956
30 pg/mL
ELISA
Colorimetric Microplate Reader
TNFRSF14
<10%
HRP
RUO
2°C to 8°C
1 hr 20 min
32.77 to 8000 pg/mL
Biotin
Plasma, Serum, Supernatant
8764
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL
TNFRSF14 (HVEM)
4 hr 45 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers