Learn More
Mettler Toledo™ Halogen Moisture Analyzer HX204
High Performance Moisture Analysis
Brand:  Mettler Toledo™ 30019875
Product Code. 12743275
  • £17585.00 / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Highly reliable and repeatable moisture results thanks to fast halogen heating and superior weighing performance
  • Define multiple users with individual rights, each with their own home screen and method shortcuts
  • Integrated routine testing features ensure the instrument is always performing optimally: FACT, SmartCal and tests for the heating and weighing units
  • Large 7 in. color touchscreen offers an easy to navigate user interface
  • Hanging weighing pan and the flat stainless steel surface underneath allow fast and easy cleaning
  • For easy reporting, connect the instrument to a PC, FTP server, strip printer or network printer using either Wi-Fi, Ethernet, RS232 or USB
TRUSTED_SUSTAINABILITY
Specifications
Specifications
115 V or 230 V
Drying