Learn More
HYAL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556152
Detalles adicionales : Weight : 0.02000kg
Descripción
Immunogen sequence: NNGRCIRKMW NAPSYLHLNP ASYHIEASED GEFTVKGKAS DTDLAVMADT FSCHCYQGYE GADCREIKTA DGCSGVSPSP GSLMTLCLLL LASYRSIQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.
Endo-hyaluronidase that degrades hyaluronan to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.Especificaciones
HYAL4 | |
Polyclonal | |
Unconjugated | |
Hyal4 | |
4632428M18Rik; Chondroitin sulfate endo-beta-N-acetylgalactosaminidase; chondroitin sulfate hydrolase; CSHY; Hyal4; hyal-4; Hyaluronidase; hyaluronidase 4; hyaluronidase-4; hyaluronoglucosaminidase 4; hyaluronoglucosaminidase-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23553 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M3T9 | |
Hyal4 | |
Recombinant protein corresponding to Human HYAL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only