missing translation for 'onlineSavingsMsg'
Learn More
Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals Product Code.: 15480687

Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals

Product Code. 15480687
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15480687

Brand: Thermo Scientific Alfa Aesar J66993.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hyaluronic acid sodium salt, CAS # 9067-32-7, is an acidic mucopolysaccharide produced by the bacterium Streptococcus zooepidemicus that is abundant in the mammalian extracellular matrix.

Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hyaluronic acid sodium salt is a component of the extracellular matrix that is potentially useful for soft tissue augmentation procedures

Application

• Hyaluronic acid sodium salt may be used in formulations of non-animal gels used in soft tissue augmentation studies
• It can be used in laboratory development of artificial synovial fluid
• It can also be used to study its role in oxidative stress and tumor progression

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9067-32-7
Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.30
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

Type Hyaluronic Acid Sodium Salt
Physical Form Powder
Color White
Quantity 1 g
Merck Index 14,4757
Solubility Information Soluble in water
Percent Purity 91%
Chemical Name or Material Hyaluronic acid sodium salt, Streptococcus equi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.