missing translation for 'onlineSavingsMsg'
Learn More

Techne™ Hybrigene Tube Holder for HB-3D

Expand the capabilities of your incubator with these parts and accessories.

Brand:  Techne™ 39455-29

Product Code. 12046625

  • £580.00 / Each
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Provided with clips
  • Available for either 16 x 15 mL or 8 x 50 mL
  • Aids in applications such as nucleic acid hybridizations such as Northern (RNA) blots/hybridizations, Southern (DNA) blots/hybridizations, Western (protein) blots, and molecular biology assays
Specifications

Specifications

16 x 15mL Tubes
Tube Carrier/Holder
Northern, Southern, Western blots and molecular biology assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers