Learn More
Cytiva HyCell STEM™ Media
Brand: Cytiva SR30004.01
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Culture human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells with HyCell-STEM™ Media, a complete media intended for use with stem cells on feeder layer cultures. This defined, serum-free formulation is used for expanding and maintaining stem cell colonies on feeder layer cultures with daily or every other day feeds, resulting in increased cell growth with fewer passages, all while maintaining pluripotency.
Specifications
HyCell-STEM-FF 6x supplement | |
HyCell-STEM-FF 6x Supplement | |
500mL | |
Culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells |