Learn More
Cytiva HyClone™ 100mM Sodium Pyruvate Solution
Filter sterilized. HyClone™ 100mM Sodium Pyruvate solution is an easily accessible carbohydrate source that may also have protective effects against hydrogen peroxide.
Brand: Cytiva SH30239.01
9.38 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Code : 83
Additional Details : Weight : 0.14040kg
Description
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
cGMP and ISO 91:2 Guidelines | |
Sodium Pyruvate, 100mM Solution | |
0.1 μm Sterile-filtered | |
100 mL |
2° to 8°C | |
100 mM | |
=1.0EU/mL |
Safety and Handling
- HyClone Sodium Pyruvate Solution
Hazard Statement
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN AQUATIC-Contains 1.2% of components with unknown hazards to the aquatic environment.