Learn More
Cytiva HyClone™ ActiCHO™ P Powdered Media
Hydrolysate-free and animal-derived component-free (ADCF)
Brand: Cytiva SH31025.08
53.82 GBP valid until 2024-12-31
Use promo code "21885" to get your promotional price.
Additional Details : CAS Number : 56-45-1 Weight : 0.03810kg
Description
ActiCHO P powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides, or growth factors in ActiCHO P medium.
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
No Glutamine, No Insulin |
Powder | |
Poloxamer 188 | |
1 L |