Learn More
Cytiva HyClone™ ActiPro Powdered Cell Culture Media
Formulated to provide high yields of recombinant proteins in bioprocesses using Chinese hamster ovary (CHO) cell lines.
£59.45 - £227.00
Specifications
Content And Storage | 2 to 8°C, away from light and moisture |
---|---|
Form | Powder |
Sterility | Non-sterile |
With Additives | Poloxamer 188 |
Without Additives | No Glutamine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17205906
|
Cytiva
SH31037.08 |
1 L |
£59.45
1 L |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15766398
|
Cytiva
SH31037.01 |
5 L |
£227.00
5 L |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HyClone ActiPro cell culture medium is available in both liquid and powder forms for cell culture bioprocesses. This chemically defined medium is intended for use in combination with HyClone ActiSM medium and Cell Boost 7a and Cell Boost 7b supplements.
ActiPro media and supplements are formulated for industrial production of recombinant proteins, including those requiring complex post-translational modifications and correct protein folding. In addition, this engineered ActiPro system of media and supplements can significantly reduce production process time.
- Demonstrates high yields of recombinant protein
- Does not contain hypoxanthine and thymidine (HAT supplement)
- Chemically defined ADCF mAb media formulation
- Production medium for batch or fed-batch fermentation processes
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
No Glutamine |
Powder | |
Poloxamer 188 |