Learn More
Cytiva HyClone™ ADCF-mab Powdered Media
Protein-free and animal-derived component-free medium
Brand: Cytiva SH30635.01
Code : 83
Additional Details : Weight : 0.01000kg
Description
ADCF‐mAb powder medium does not contain poloxamer 188, L-glutamine, phenol red, or sodium bicarbonate.
ADCF-mAb is available in liquid and powder formats in user-friendly packaging.
- Animal-derived component-free (ADCF) formulation
- Designed for high cell yield and monoclonal antibody (MAb) production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
No Glutamine, No Phenol Red |
Powder | |
HEPES | |
5 L |