missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ AdvanceSTEM™ ES qualified Hepes buffer
Brand: Cytiva SH30851.01
14.93 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Code : 85
Additional Details : Weight : 0.10500kg
Specifications
cGMP and ISO 91:2 Guidelines | |
AdvanceSTEM ES Qualified HEPES (1M) | |
0.1 μm Sterile-filtered | |
7.2 to 7.5 |
2° to 8°C | |
1 M | |
FIO | |
100 mL |