Learn More
Cytiva HyClone™ Bovine Calf Serum (BCS), US Origin, Irradiated 25-40 kGy
A cost-effective alternative to Fetal Bovine Serum (FBS).
Brand:  Cytiva SH30073.04IR25-40
Product Code. 17622075
  • £138.00 / 1 L
Explore more special offers
Description
Description
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16-24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
1000 mL
≤ 10 EU/mL
Not Detected
United States
Irradiated
< 30 mg/dL