missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Growth Serum

Product Code. p-8013208
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
None
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 16201122

Brand: Cytiva SH30541.03IR2540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cost-effective alternative to Fetal Bovine Serum (FBS).

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
Content And Storage <–1°C
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated (25 to 40 doses)
Tested For Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.